Ammonium CRMs, standard solutions, Supelco®
Toimittaja: Merck
1.25023.0100 1.25024.0100 1.25022.0100 1.25025.0100 1.25026.0100 1.25027.0100 1.32227.0100
1.25023.0100EA 63.1 EUR
1.25023.0100 1.25024.0100 1.25022.0100 1.25025.0100 1.25026.0100 1.25027.0100 1.32227.0100
Ammonium CRMs, standard solutions, Supelco®
Standardiaineet ja vertailumateriaalit Cationic Standards Ammonium Standards
NH₄Cl in H₂O
  • Diluted standard solutions that are ready-to-use for accurate measurements
  • Traceable to SRM from NIST and USP
  • Full certificates of analysis with comprehensive quality documentation
  • Information on concentration and expanded measurement uncertainty
  • Save time and minimize errors
Method used for Uncertainty Measurement (Accuracy in the table below)= Photometry
Sertifikaatit: Traceable to SRM from NIST and USP.