Extraction unit and drying attachment, LiChrolut®
Toimittaja: Merck
1.19851.0001 1.19852.0001 1.19874.0001
1.19851.0001EA 2400 EUR
1.19851.0001 1.19852.0001 1.19874.0001
Extraction unit and drying attachment, LiChrolut®
Uuttolaitteet ja uutoshylsyt Soxhlet- ja Soxhlet-tyypin uuttolaitteet
All the individual steps associated with solid phase extraction can be carried out using the LiChrolut® extraction unit. This transparent and vacuum-suitable unit, made of glass, can be used to prepare up to 12 samples simultaneously.
  • Control of the vacuum via a manometer located at the front
  • Individual, easy setting of the various flow rates using valves
Toimitustiedot: Supplied with 1 lid with 12 standard valves and seal, 1 glass chamber with gauge and vacuum valve, 12 standard stainless steel cannules, 1 collecting rack (base plate with 3 supportMore Product Information