Tetrabutyyliammoniumvetysulfaatti, LiChropur™ for ion pair chromatography, Supelco®

Toimittaja: Merck
Varoitus


M-Clarity™ quality level = MQ100

1.18312.0025 1.18312.0100
1.18312.0025EA 623 EUR
1.18312.0025 1.18312.0100
Tetrabutyyliammoniumvetysulfaatti, LiChropur™ for ion pair chromatography, Supelco®
Tetrabutyyliammoniumvetysulfaatti
Kaava: (C₄H₉)₄NHSO₄
Sulamispiste: 169…172 °C
Tiheys: 1,01 g/cm³ (25 °C)
MDL numero: MFCD00011637
CAS-numero: 32503-27-8
EINECS: 251-068-5

Limited Time Sale

Select items below are currently on sale

Order Now

Testitulosten erittely

Assay (acidimetric; calculated on dried substance) ≥ 99%
Identity (IR-spectrum) passes test
pH-value (10 %; water) ≤ 2
UV-transmission (200 nm; 0.005 mol/l; 1 cm; water) ≥ 70 %
UV-transmission (220 nm; 0.005 mol/l; 1 cm; water) ≥ 90 %
UV-transmission (250 nm; 0.005 mol/l; 1 cm; water) ≥ 98 %
Loss on drying (120 °C, vacuum) ≤ 2%

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR