Reflectoquant®-testiliuskat, Supelco®
Toimittaja: Merck
1.16128.0001
1.16128.0001EA 148 EUR
1.16128.0001
Reflectoquant®-testiliuskat, Supelco®
Ympäristöanalytiikan pikatestit
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
  • Fast analysis with reliable results
  • Barcode calibration
  • Low analysis costs
  • Environmentally friendly