Asetonitriili, LiChrosolv®, isokraattinen laatu for liquid chromatography, Supelco®

Toimittaja: Merck
Vaara


M-Clarity™ quality level = MQ100

1.14291.1000 1.14291.2500 1.14291.4000 1.14291.9010 1.14291.9030 1.14291.9185 1.14291.9910
1.14291.1000EA 105 EUR
1.14291.1000 1.14291.2500 1.14291.4000 1.14291.9010 1.14291.9030 1.14291.9185 1.14291.9910
Asetonitriili, LiChrosolv®, isokraattinen laatu for liquid chromatography, Supelco®
Asetonitriili

With their high degree of UV transmittance, low particle count, low acidity and alkalinity and low evaporation residue level, LiChrosolv® solvents are ideal for reproducible separations. Since separations are normally carried out under gradient conditions in analytical HPLC, we offer solvents in "gradient grade" as well as "isocratic grade". This enables to minimize the gradient effect of the solvent involved.

Kaava: H₃CCN
Kiehumispiste: 81,6 °C (1013 hPa)
Sulamispiste: –45,7 °C
Tiheys: 0,782 g/cm³ (20 °C)
Leimahduspiste: 2 °C (sulj. astia)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00001878
CAS-numero: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR