Kloorivetyliuos 0.357 mol/l (N/2.8), Titripur® volumetric solution, Supelco®

Toimittaja: Merck
Varoitus


M-Clarity™ quality level = MQ100

1.13136.9010
1.13136.9010EA 175 EUR
1.13136.9010
Kloorivetyliuos 0.357 mol/l (N/2.8), Titripur® volumetric solution, Supelco®
Kloorivetyliuos

Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.


Stability of product in Titripac packaging: Merck have carried out testing to show that products are stable for 36 months when in use and opened. Please see Technical report (Product specification) in the Related Information tab below.

Kaava: HCl
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011324
CAS-numero: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR