Hopeanitraatti 0.1 mol/l concentrated aqueous solution, Titrisol®, Supelco®

Toimittaja: Merck
Vaara


M-Clarity™ quality level = MQ100

1.09990.0001
1.09990.0001EA 99.9 EUR
1.09990.0001
Hopeanitraatti 0.1 mol/l concentrated aqueous solution, Titrisol®, Supelco®
Hopeanitraatti
Kaava: AgNO₃
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003414
CAS-numero: 7761-88-8
EINECS: 231-853-9
UN: 1760
ADR: 8,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Solubility (20 °C) soluble
Density 1.27 g/cm3 (20 °C)
pH 7 - 9 (H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR