Butyyliasetaatti ≥99.5% (by GC), EMSURE® for analysis, Supelco®

Toimittaja: Merck
Varoitus


M-Clarity™ quality level = MQ300

1.09652.1000 1.09652.2500 1.09652.4000
1.09652.1000EA 91.4 EUR
1.09652.1000 1.09652.2500 1.09652.4000
Butyyliasetaatti ≥99.5% (by GC), EMSURE® for analysis, Supelco®
Butyyliasetaatti

For full details of specification and hazards please see www.vwr.com or the VWR Chemicals catalogues

Kaava: CH₃COO(CH₂)₃CH₃
Kiehumispiste: 126 °C (1013 hPa)
Sulamispiste: –76 °C
Tiheys: 0,875 g/cm³ (20 °C)
Leimahduspiste: 25 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00009445
CAS-numero: 123-86-4
EINECS: 204-658-1
UN: 1123
ADR: 3,
Merck Index: 13,01534
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=403329de943f8cb2ea6cd9d73ee35957d2140aa7&vid=682ab82de177b2985e94089408f7efebde59926d&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Purity (GC) ≥ 99.5 %
Identity (IR) conforms
Color ≤ 10 Hazen
Acidity ≤ 0.0017 meq/g
Readily carbonizable substances conforms
n-Butyl alcohol (GC) ≤ 0.2 %
n-Butyl formiate (GC) ≤ 0.1 %
n-Butyl propionate (GC) ≤ 0.1 %
Evaporation residue ≤ 0.001 %
Water ≤ 0.1 %
Al (Aluminium) ≤ 0.00005 %
B (Boron) ≤ 0.000002 %
Ba (Barium) ≤ 0.00001 %
Ca (Calcium) ≤ 0.00005 %
Cd (Cadmium) ≤ 0.000005 %
Co (Cobalt) ≤ 0.000002 %
Cr (Chromium) ≤ 0.000002 %
Cu (Copper) ≤ 0.000002 %
Fe (Iron) ≤ 0.00001 %
Mg (Magnesium) ≤ 0.00001 %
Mn (Manganese) ≤ 0.000002 %
Ni (Nickel) ≤ 0.000002 %
Pb (Lead) ≤ 0.00001 %
Sn (Tin) ≤ 0.00001 %
Zn (Zinc) ≤ 0.00001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR