pH Indicator strips, special indicator for meat processing industry, MQuant®, Supelco®
Toimittaja: Merck
1.09632.0001
1.09632.0001EA 20.6 EUR
1.09632.0001
pH Indicator strips, special indicator for meat processing industry, MQuant®, Supelco®
Testipaperit, pH-liuskat pH-testiliuskat
The measurement of pH in meat plays a central role in the processing of meat products and the assessment of their quality.
  • No 'contamination' of the measurement site thanks to the covalent binding of the indicator dyes
  • The test strip principle enables the measurement to be made directly in the meat (the plastic material is physiologically safe)
  • No protein error
The slaughtering procedure triggers a series of biochemical processes that result in a shift of the initial value (pH 7,0) into the acidic range (pH 5,4 to 6,7). These strips were specificallyMore Product Information