Kaliumhydroksidi 0.1 mol/l (0.1 N) in ethanol, Titripur® Reag. USP, Reag. Ph. Eur. volumetric solution, Supelco®

Toimittaja: Merck
Vaara

Synonyms: Kalilipeä

M-Clarity™ quality level = MQ100

1.09115.1000 1.09115.2500 1.09115.1003
1.09115.1000EA 79.7 EUR
1.09115.1000 1.09115.2500 1.09115.1003
Kaliumhydroksidi 0.1 mol/l (0.1 N) in ethanol, Titripur® Reag. USP, Reag. Ph. Eur. volumetric solution, Supelco®
Kaliumhydroksidi

Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.

Kaava: KOH
Tiheys: 0,836 g/cm³ (20 °C)
Leimahduspiste: 12 °C (EtOH)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003553
CAS-numero: 1310-58-3
EINECS: 215-181-3
UN: 2924
ADR: 3 (8),II

Order Now

Testitulosten erittely

Form liquid
Amount-of-substance concentration 0.0995-0.1005
Measurement uncertainty +/- 0.0003
Traceability NIST SRM


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR