Kloorivetyliuos 0,1 mol/l (0,1 N), Titripur® Reag. Ph. Eur., Reag. USP mittaliuos, Supelco®

Toimittaja: Merck
Varoitus


M-Clarity™ quality level = MQ100

1.09060.1000 1.09060.5000 1.09060.9010 1.09060.9025 1.09060.4000 1.09060.1003 1.09060.4003 1.09060.9013
1.09060.1000EA 36.9 EUR
1.09060.1000 1.09060.5000 1.09060.9010 1.09060.9025 1.09060.4000 1.09060.1003 1.09060.4003 1.09060.9013
Kloorivetyliuos 0,1 mol/l (0,1 N), Titripur® Reag. Ph. Eur., Reag. USP mittaliuos, Supelco®
Kloorivetyliuos

Merck have introduced into their Titripur® Titrants range packs with additional technology - Supelco® SmartTitrants use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. Mettler Toledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.


Stability of product in Titripac packaging: Merck have carried out testing to show that products are stable for 36 months when in use and opened. Please see Technical report (Product specification) in the Related Information tab below.

Kaava: HCl
Tiheys: 1,004 g/cm³ (25 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011324
CAS-numero: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Order Now

Testitulosten erittely

Form liquid
Amount-of-substance concentration 0.0995-0.1005
Measurement uncertainty +/- 0.0003
Traceability NIST SRM


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR