Sinkki ≥99.9%, sticks, EMSURE® for analysis, Supelco®

Toimittaja: Merck


M-Clarity™ quality level = MQ300

1.08782.0500
1.08782.0500EA 156 EUR
1.08782.0500
Sinkki ≥99.9%, sticks, EMSURE® for analysis, Supelco®
Sinkki

Sticks, triangular cross section about 8 mm

Kaava: Zn
Kiehumispiste: 908 °C (1013 hPa)
Sulamispiste: 420 °C
Tiheys: 7,14 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011291
CAS-numero: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255

Order Now

Testitulosten erittely

Assay (complexometric) ≥ 99.9 %
As (Arsenic) ≤ 0.00001 %
Cd (Cadmium) ≤ 0.005 %
Cu (Copper) ≤ 0.002 %
Fe (Iron) ≤ 0.005 %
Pb (Lead) ≤ 0.005 %
Sn (Tin) ≤ 0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR