TRITON™ X-100 analytical reagent, Millipore®

Toimittaja: Merck
Vaara

Synonyms: Polyethyleneglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]-ether, (p-tert-Octylphenoxy)polyethoxyethanol, Octoxynol, Polyoxyethylene(10) Octylphenyl Ether, t-Octylphenoxypolyethoxyethanol, O-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanol

M-Clarity™ quality level = MQ200

1.08603.1000 1.08603.2500 1.08603.6025
1.08603.1000EA 125 EUR
1.08603.1000 1.08603.2500 1.08603.6025
TRITON™ X-100 analytical reagent, Millipore®
TRITON™ X-100
Kiehumispiste: >200 °C (1013 hPa)
Sulamispiste: 6 °C
Tiheys: 1,07 g/cm³ (20 °C)
Leimahduspiste: 251 °C (sulj. astia)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00132505
CAS-numero: 9036-19-5
UN: 3082
ADR: 9,III
Merck Index: 13,06793

Order Now

Testitulosten erittely

Identity (IR spectrum) passes test
pH value (10 g/l, H₂O, 20 °C) 5.0 - 8.0
Density (d 20/4) 1.064 - 1.067
Cloud point 63 - 69 °C
Ethylene oxide (GC) ≤30 ppm
Dioxane (GC) ≤20 ppm
Sulphated ash (600 °C) ≤0.2%
Water (acc. to Karl Fischer) ≤0.2%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR