Tolueeni, SupraSolv® for GC-ECD/FID, Supelco®

Toimittaja: Merck
Vaara

Synonyms: Methylbenzene, Phenylmethane, FM approval

M-Clarity™ quality level = MQ100

1.08389.1000 1.08389.2500 1.08389.4000
1.08389.1000EA 79.5 EUR
1.08389.1000 1.08389.2500 1.08389.4000
Tolueeni, SupraSolv® for GC-ECD/FID, Supelco®
Tolueeni

SupraSolv® ECD and FID is specially developed for gas chromatography in combination with ECD (Electron Capture Detetor) and FID (Flame Ionization detector). SupraSolv® solvents offer the largest specified retention time range, a clear baseline and a minimal signal-to-noise ratio leading to reliable and reproducible analysis results.

Kaava: C₆H₅CH₃
Kiehumispiste: 110,6 °C (1013 hPa)
Sulamispiste: –95 °C
Tiheys: 0,867 g/cm³ (20 °C)
Leimahduspiste: 4 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00008512
CAS-numero: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3,II
REACH: 05-2114615130-69
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7a09f4eae3dd5e25d737baeeb21ad0c7e8f13bd&vid=1e05dbed32d28a28b5ae333c7d1d140108a340f0&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Solubility 0.52 g/l (20 °C)
Melting Point -95 °C
Molar Mass 92.14 g/mol
Boiling Point 110.6 °C (1013 hPa)
Vapor Pressure 29 hPa (20 °C)
Flash Point 4 °C
Refractive Index 1.4968 (20 °C)
Explosion Limit 1.2 - 8 %(V)
Density 0.87 g/cm3 (20 °C)
pH (H2O) not applicable
Ignition Point 535 °C
Viscosity kinematic 0.7 mm2/s (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR