n-Pentaani ∼ 95%, EMPLURA®, Supelco®

Toimittaja: Merck
Vaara


M-Clarity™ quality level = MQ200

1.07176.1000 1.07176.9190 1.07176.2500
1.07176.1000EA 82.8 EUR
1.07176.1000 1.07176.9190 1.07176.2500
n-Pentaani ∼ 95%, EMPLURA®, Supelco®
n-Pentaani
Kaava: H₃C(CH₂)₃CH₃
Kiehumispiste: 35…36 °C (1013 hPa)
Sulamispiste: –130 °C
Tiheys: 0,63 g/cm³ (20 °C)
Leimahduspiste: –40 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00009498
CAS-numero: 109-66-0
EINECS: 203-692-4
UN: 1265
ADR: 3,II
REACH: 01-2119459286-30
Merck Index: 13,07193
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ba2c9c81a4b1f44769627a00ba49b6cc8e63132&vid=f6831eb15ab1602d8c752468dc3ee31cc4ee61c0&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Purity (GC) ≥ 95.0 %
Identity (IR) conforms
Free acid (as CH₃COOH) ≤ 0.001 %
Density (d 20 °C/ 4 °C) 0.626 - 0.628
Readily carbonizable substances conforms
Evaporation residue ≤ 0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR