L(+)-dinatriumtartraatti-2 H2O, EMSURE® for analysis, Supelco®

Toimittaja: Merck


M-Clarity™ quality level = MQ300

1.06663.0250 1.06663.1000 1.06663.9050
1.06663.0250EA 94.9 EUR
1.06663.0250 1.06663.1000 1.06663.9050
L(+)-dinatriumtartraatti-2 H2O, EMSURE® for analysis, Supelco®
L(+)-dinatriumtartraatti-2 H2O
Kaava: NaOOC(CHOH)₂COONa·2H₂O
Sulamispiste: 57 °C
Tiheys: 1,818 g/cm³ (20 °C)
MDL numero: MFCD00150035
CAS-numero: 6106-24-7
EINECS: 212-773-3
Merck Index: 12,08833
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39a75038f4ff5e6c234267b15540823ae361bed4&vid=89289572c48b0c6415f8f11e916a51f66e129474&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Solubility 290 g/l (20 °C)
Melting Point 57 °C
Molar Mass 230.08 g/mol
Bulk Density 460 kg/m3
Density 1.82 g/cm3
pH 8 (50 g/l, H2O, 25 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR