Trinatriumfosfaatti-12 H2O, EMSURE® Reag. Ph. Eur., ACS for analysis, Supelco®

Toimittaja: Merck
Varoitus

Synonyms: Sodium Phosphate tribasic 12-Hydrate, Sodium phosphate dodecahydrate, Sodium phosphate tribasic dodecahydrate

M-Clarity™ quality level = MQ300

1.06578.1000 1.06578.5000 1.06578.9050 1.06578.9012
1.06578.1000EA 46.5 EUR
1.06578.1000 1.06578.5000 1.06578.9050 1.06578.9012
Trinatriumfosfaatti-12 H2O, EMSURE® Reag. Ph. Eur., ACS for analysis, Supelco®
Trinatriumfosfaatti-12 H2O
Kaava: Na₃PO₄·12H₂O
Sulamispiste: ∼75 °C
Tiheys: 1,62 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149198
CAS-numero: 10101-89-0
EINECS: 231-509-8
ADR: 8,III
REACH: 01-2119489800-32-xxxx
Merck Index: 13,08736

Order Now

Testitulosten erittely

Solubility 285 g/l (20 °C)
Melting Point 75 °C Elimination of water of crystallisation
Molar Mass 380.18 g/mol
Bulk Density 950 kg/m3
Density 1.62 g/cm3 (20 °C)
pH 12 (10 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR