Natriumvetykarbonaatti 99.0-101.0%, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, USP, FCC, E500, SAFC®

Toimittaja: MERCK PRODUCTION CHEMICALS

1.06323.2500 1.06323.9029 1.06323.9500
1.06323.2500EA 187 EUR
1.06323.2500 1.06323.9029 1.06323.9500
Natriumvetykarbonaatti 99.0-101.0%, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, USP, FCC, E500, SAFC®
Natriumvetykarbonaatti

Residues of metal catalysts or metal reagents acc. to EMEA/CHMP/SWP/4446/2000 are not likely to be present.
Corresponds to Ph Eur, BP, USP, FCC, JP, E 500
Conforms to the purity criteria on food additives according to the current European Commission Regulation.

Kaava: NaHCO₃
Sulamispiste: 270 °C
Tiheys: 2,159 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003528
CAS-numero: 144-55-8
EINECS: 205-633-8
Merck Index: 13,08655
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Assay (acidimetric; calculated on dried substance) 99.5 - 100.5 %
Assay (acidimetric) 99.0 - 101.0 %
Identity passes test
Appearance of solution passes test
Insoluble matter passes test
pH-value (1 %, water) 8.0 - 8.6
pH-value (5 %; water) 7.9 - 8.4
Carbonate (CO₃) passes test
Chloride (Cl) ≤ 0.015 %
Sulphate (SO₄) ≤ 0.01 %
Sulfur compounds (as SO₄) ≤ 0.015 %
Heavy metals (as Pb) ≤ 0.0005 %
Al (Aluminium) ≤ 0.00005 %
As (Arsenic) ≤ 0.0002 %
Ca (Calcium) ≤ 0.01 %
Cu (Copper) ≤ 0.0005 %
Fe (Iron) ≤ 0.002 %
Hg (Mercury) ≤ 0.0001 %
NH₄ (Ammonium) ≤ 0.002 %
NH₄ (Ammonium) according to USP ≤ 0.002 %
NH₄ (Ammonium) according to JP passes test
Pb (Lead) ≤ 0.0002 %
Zn (Zinc) ≤ 0.0025 %
Residual solvents (ICH Q3C) excluded by production process
Loss on drying (Silica gel) ≤ 0.25 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR