Metanoli ≥99.9%, EMSURE® Reag. Ph. Eur., ACS, ISO for analysis, Supelco®

Toimittaja: Merck
Vaara

Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol

M-Clarity™ quality level = MQ300

1.06009.1000 1.06009.1011 1.06009.2500 1.06009.2511 1.06009.5000 1.06009.6010 1.06009.6025 1.06009.6190 1.06009.9025 1.06009.9180 1.06009.9450 1.06009.9900 1.06009.9999 1.06009.9950 1.06009.9905 1.06009.4000
1.06009.1000EA 14.67 EUR
1.06009.1000 1.06009.1011 1.06009.2500 1.06009.2511 1.06009.5000 1.06009.6010 1.06009.6025 1.06009.6190 1.06009.9025 1.06009.9180 1.06009.9450 1.06009.9900 MERC1.06009.9999 1.06009.9950 1.06009.9905 1.06009.4000
Metanoli ≥99.9%, EMSURE® Reag. Ph. Eur., ACS, ISO for analysis, Supelco®
Metanoli

EMSURE® grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE® provides worldwide best and most extensive product specifications. We declare our EMSURE® range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.

Kaava: CH₃OH
Kiehumispiste: 64,5 °C (1013 hPa)
Sulamispiste: –98 °C
Tiheys: 0,792 g/cm3 (20 °C)
Leimahduspiste: 9,7 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00004595
CAS-numero: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3 (6.1),II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Purity (GC) ≥ 99.9 %
Identity (IR) conforms
Appearance clear
Color ≤ 10 Hazen
Solubility in water conforms
Acidity ≤ 0.0002 meq/g
Alkalinity ≤ 0.0002 meq/g
Density (d 20 °C/20 °C) 0.791 - 0.793
Boiling point 64 - 65 °C
Benzene (impurity A) (GC) ≤ 2 ppm
Ethanol (GC) ≤ 0.05 %
Acetone ≤ 0.001 %
Acetaldehyde ≤ 0.001 %
Formaldehyde ≤ 0.001 %
Readily carbonizable substances conforms
Carbonyl compounds (as CO) ≤ 0.001 %
Chloride (Cl) ≤ 0.5 ppm
Sulfate (SO₄) ≤ 1 ppm
Substances reducing potassium permanganate (as O) ≤ 0.00025 %
Ag (Silver) ≤ 0.000002 %
Al (Aluminium) ≤ 0.00005 %
As (Arsenic) ≤ 0.000002 %
Au (Gold) ≤ 0.000002 %
Ba (Barium) ≤ 0.00001 %
Be (Beryllium) ≤ 0.000002 %
Bi (Bismuth) ≤ 0.000002 %
Ca (Calcium) ≤ 0.00005 %
Cd (Cadmium) ≤ 0.000005 %
Co (Cobalt) ≤ 0.000002 %
Cr (Chromium) ≤ 0.000002 %
Cu (Copper) ≤ 0.000002 %
Fe (Iron) ≤ 0.00001 %
Ga (Gallium) ≤ 0.000002 %
In (Indium) ≤ 0.000002 %
Li (Lithium) ≤ 0.000002 %
Mg (Magnesium) ≤ 0.00001 %
Mn (Manganese) ≤ 0.000002 %
Mo (Molybdenum) ≤ 0.000002 %
Ni (Nickel) ≤ 0.000002 %
Pb (Lead) ≤ 0.00001 %
Pt (Platinum) ≤ 0.000005 %
Sn (Tin) ≤ 0.00001 %
Ti (Titanium) ≤ 0.000002 %
Tl (Thallium) ≤ 0.000002 %
V (Vanadium) ≤ 0.000002 %
Zn (Zinc) ≤ 0.00001 %
Zr (Zirconium) ≤ 0.000002 %
Evaporation residue ≤ 0.0005 %
Water ≤ 0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR