Magnesiumperkloraatti-H2O elementary_analysis, Supelco®

Toimittaja: Merck
Vaara

Synonyms: Anhydrone hydrate, Magnesium perchlorate hydrat

MQ100

1.05875.0500
1.05875.0500EA 338 EUR
1.05875.0500
Magnesiumperkloraatti-H2O elementary_analysis, Supelco®
Magnesiumperkloraatti-H2O
Kaava: Cl₂MgO₈·H₂O
MW: 241.22 g/mol
Sulamispiste: 146
Tiheys: 2.6 g/cm³ (25 °C) (anhydr.)
Storage Temperature: Huoneenlämpö
CAS-numero: 64010-42-0
EINECS: 233-108-3
UN: 1475
ADR: 5.1 II

Order Now

Testitulosten erittely

Solubility 500 g/l (25 °C)
Melting Point 251 °C decomposes
Bulk Density 600 - 800 kg/m3
Density 2.60 g/cm3 (25 °C) (anhydrous substance)
pH 8.2 (50 g/l, H2O, 25 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR