Magnesiumasetaatti-4 H2O, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®

Toimittaja: Merck


M-Clarity™ quality level = MQ300

1.05819.0250 1.05819.1000 1.05819.9050
1.05819.0250EA 109 EUR
1.05819.0250 1.05819.1000 1.05819.9050
Magnesiumasetaatti-4 H2O, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Magnesiumasetaatti-4 H2O
Kaava: (H₃CCOO)₂Mg·4H₂O
Sulamispiste: 80 °C
Tiheys: 1,454 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149214
CAS-numero: 16674-78-5
EINECS: 205-554-9
Merck Index: 13,05676

Order Now

Testitulosten erittely

Solubility 1200 g/l (15 °C)
Melting Point 80 °C
Molar Mass 214.46 g/mol
Bulk Density 510 kg/m3
Density 1.45 g/cm3
pH 6.1 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR