Molekyyliseula A4, (0,4 nm, 4 Å), beads Reag. Ph. Eur. 2 mm, Supelco®

Toimittaja: Merck


M-Clarity™ quality level = MQ200

1.05708.0250 1.05708.1000 1.05708.9010
1.05708.0250EA 126 EUR
1.05708.0250 1.05708.1000 1.05708.9010
Molekyyliseula A4, (0,4 nm, 4 Å), beads Reag. Ph. Eur. 2 mm, Supelco®
Molekyyliseula A4, (0,4 nm, 4 Å)
Tiheys: 1,1…2.2 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD03457537
CAS-numero: 70955-01-0
EINECS: 215-283-8

Order Now

Testitulosten erittely

Solubility (20 °C) insoluble
Bulk Density 700 - 750 kg/m3
Grade Reag. Ph Eur
Synonyms Sodium aluminium silicate


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR