L(+)-lysiinimonohydrokloridi, Millipore®

Toimittaja: Merck


M-Clarity™ quality level = MQ100

1.05700.1000 1.05700.9010
1.05700.1000EA 219 EUR
1.05700.1000 1.05700.9010
L(+)-lysiinimonohydrokloridi, Millipore®
L(+)-lysiinimonohydrokloridi
Kaava: H₂N(CH₂)₄CH(NH₂)CO₂H·HCl
Sulamispiste: 263…264 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00064564
CAS-numero: 657-27-2
EINECS: 211-519-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR