TLC-levyt, TLC- ja PLC-levyt, alumiinioksidi 60
Toimittaja: Merck
1.05550.0001 1.05581.0001 1.05713.0001 1.05731.0001
1.05550.0001EA 318 EUR
1.05550.0001 1.05581.0001 1.05713.0001 1.05731.0001
TLC-levyt, TLC- ja PLC-levyt, alumiinioksidi 60
TLC-levyt
Merck TLC aluminium oxide plates use neutral or basic aluminium oxide of 60 or 150 Å pore size with or without fluorescent indicator to suit different application needs. Under aqueous conditions, basic compounds can be best separated on basic aluminium oxide plates, while neutral compounds are best separated on neutral plates.
  • Erilaiset erottelut eri pH-alueilla