Ammoniakkiliuos 25%, EMSURE® Reag. Ph. Eur., ISO for analysis, Supelco®

Toimittaja: Merck
Vaara

Synonyms: Ammoniakki

M-Clarity™ quality level = MQ300

1.05432.1000 1.05432.1011 1.05432.2500 1.05432.5000 1.05432.9025 1.05432.9181 1.05432.2511
1.05432.1000EA 40.7 EUR
1.05432.1000 1.05432.1011 1.05432.2500 1.05432.5000 1.05432.9025 1.05432.9181 1.05432.2511
Ammoniakkiliuos 25%, EMSURE® Reag. Ph. Eur., ISO for analysis, Supelco®
Ammoniakkiliuos
Kaava: NH₃
Kiehumispiste: 32…37,7 °C (1013 hPa)
Sulamispiste: –60…–57,5 °C
Tiheys: 0,88…0.91 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00066650
CAS-numero: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8,III

Order Now

Testitulosten erittely

Assay (acidimetric, NH₃) 25.0 - 30.0 %
Identity passes test
Appearance of solution passes test
Density (20/20°C) 0.892 - 0.910
Carbonate (as CO₂) ≤ 10 ppm
Chloride (Cl) ≤ 0.5 ppm
Phosphate (PO₄) ≤ 0.5 ppm
Silicon (as SiO₂) ≤ 10 ppm
Sulfate (SO₄) ≤ 2 ppm
Sulphide (S) ≤ 0.2 ppm
Pyridine and related substances ≤ 2 ppm
Ag (Silver) ≤ 0.020 ppm
Al (Aluminium) ≤ 0.500 ppm
Au (Gold) ≤ 0.100 ppm
Ba (Barium) ≤ 0.050 ppm
Bi (Bismuth) ≤ 0.100 ppm
Ca (Calcium) ≤ 0.500 ppm
Cd (Cadmium) ≤ 0.050 ppm
Co (Cobalt) ≤ 0.050 ppm
Cr (Chromium) ≤ 0.050 ppm
Cu (Copper) ≤ 0.100 ppm
Fe (Iron) ≤ 0.100 ppm
Ga (Gallium) ≤ 0.020 ppm
In (Indium) ≤ 0.020 ppm
K (Potassium) ≤ 0.500 ppm
Li (Lithium) ≤ 0.020 ppm
Mg (Magnesium) ≤ 0.100 ppm
Mn (Manganese) ≤ 0.050 ppm
Mo (Molybdenum) ≤ 0.050 ppm
Na (Sodium) ≤ 0.500 ppm
Ni (Nickel) ≤ 0.050 ppm
Pb (Lead) ≤ 0.050 ppm
Pt (Platinum) ≤ 0.100 ppm
Sn (Tin) ≤ 0.100 ppm
Sr (Strontium) ≤ 0.100 ppm
Ti (Titanium) ≤ 0.100 ppm
Tl (Thallium) ≤ 0.050 ppm
Zn (Zinc) ≤ 0.100 ppm
Oxidizable substances passes test
Substances reducing potassium permanganate (as O) ≤ 5 ppm
Residue on ignition (as SO₄) ≤ 10 ppm
Non volatile matter ≤ 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR