D(-)-fruktoosi, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, FCC, SAFC®

Toimittaja: MERCK PRODUCTION CHEMICALS

1.05321.9525 1.05321.9029
1.05321.9525EA 0 EUR
1.05321.9525 1.05321.9029
D(-)-fruktoosi, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, FCC, SAFC®
D(-)-fruktoosi

Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).

Kaava: C₆H₁₂O₆
Kiehumispiste: 552 °C (1013 hPa)
Sulamispiste: 100…104 °C (haj. lämpötila)
Tiheys: 1,59 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00148910
CAS-numero: 57-48-7
EINECS: 200-333-3
Merck Index: 13,04295
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8c96c34b23753d10b06108938876644c747995c&vid=6d952afc3154d3befcf243a015f513a9130c0835&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Assay (α 20/D, 100 g/l, water, calc. on anhydrous substance) 98.5 - 101.2 %
Assay (α 25/D, 100 g/l,water, calc. on anhydrous substance) 98.0 - 102.0 %
Identity passes test
Appearance of solution (EP) passes test
Appearance of solution (USP) passes test
Acidity or alkalinity passes test
Spec. rotation (α 20/D; 10 %; water; calc. on anhydrous substance) -93.5 - -91.0 °
Spec. rotation (α 25/D; 10 %; water; calc. on anhydrous substance) -90.8 - -87.2 °
Chloride (Cl) ≤ 0.01 %
Sulfate (SO₄) ≤ 0.02 %
Heavy metals (as Pb) ≤ 0.0005 %
As (Arsenic) ≤ 0.0001 %
Ba (Barium) passes test
Calcium and Magnesium (as Ca) ≤ 0.005 %
Cu (Copper) ≤ 0.0025 %
Pb (Lead) ≤ 0.00001 %
Zn (Zinc) ≤ 0.0025 %
Glucose (HPLC) ≤ 0.5 %
Foreign sugars passes test
5-Hydroxymethylfurfural and related substances passes test
Residual solvents (ICH Q3C) excluded by the production process
Sulfated ash ≤ 0.1 %
Water ≤ 0.5 %
Corresponds to Ph Eur, BP, USP, FCC


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR