Kaliumpermanganaatti 99.0-100.5% (by manganometry), EMSURE® Reag. Ph. Eur., ACS for analysis, Supelco®

Toimittaja: Merck
Vaara

Synonyms: Permanganic acid potassium salt

M-Clarity™ quality level = MQ300

1.05082.0250 1.05082.1000 1.05082.0250_N 1.05082.1000_N
1.05082.0250EA 63 EUR
1.05082.0250 1.05082.1000 MERC1.05082.0250_N MERC1.05082.1000_N
Kaliumpermanganaatti 99.0-100.5% (by manganometry), EMSURE® Reag. Ph. Eur., ACS for analysis, Supelco®
Kaliumpermanganaatti

For full details of specification and hazards please see www.vwr.com or the VWR Chemicals catalogues

Kaava: KMnO₄
Sulamispiste: > 240 °C
Tiheys: 2,7 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011364
CAS-numero: 7722-64-7
EINECS: 231-760-3
UN: 1490
ADR: 5.1,II
Merck Index: 12,07824

Order Now

Testitulosten erittely

Assay (manganometric) 99.0 - 100.5 %
Identity Passes test
Appearance of solution Passes test
In water insoluble matter ≤ 0.2 %
Chloride (Cl) ≤ 0.02 %
Chloride, Chlorate (as Cl) ≤ 0.005 %
Sulfate (SO₄) ≤ 0.01 %
Total nitrogen (N) ≤ 0.003 %
Cu (Copper) ≤ 0.001 %
Fe (Iron) ≤ 0.002 %
Pb (Lead) ≤ 0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR