Dikaliumoksalaatti-H2O, EMSURE® ACS for analysis, Supelco®

Toimittaja: Merck
Varoitus


M-Clarity™ quality level = MQ300

1.05073.0250 1.05073.1000
1.05073.0250EA 62.5 EUR
1.05073.0250 1.05073.1000
Dikaliumoksalaatti-H2O, EMSURE® ACS for analysis, Supelco®
Dikaliumoksalaatti-H2O
Kaava: (COOK)₂·H₂O
Sulamispiste: 356 °C
Tiheys: 2,154 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00150033
CAS-numero: 6487-48-5
EINECS: 209-506-8
UN: 2811
ADR: 6.1,III
Merck Index: 12,07820

Order Now

Testitulosten erittely

Solubility 360 g/l (20 °C) (anhydrous substance)
Molar Mass 184.24 g/mol
Bulk Density 700 - 1100 kg/m3
Vapor Pressure (20 °C)
Density 2.13 g/cm3 (20 °C)
pH 7.0 - 8.5 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR