Kaliumdivetyortofosfaatti, EMSURE® ISO for analysis, Supelco®

Toimittaja: Merck

Synonyms: Monopotassiumphosphate, prim.-Potassium phosphate, Monopotassium phosphate, Potassium phosphate monobasic, Monobasic Potassium Phosphate, Potassium dihydrogen orthophosphate

M-Clarity™ quality level = MQ300

1.04873.0250 1.04873.1000 1.04873.5000 1.04873.9025 1.04873.9050
1.04873.0250EA 28.56 EUR
1.04873.0250 1.04873.1000 1.04873.5000 1.04873.9025 1.04873.9050
Kaliumdivetyortofosfaatti, EMSURE® ISO for analysis, Supelco®
Kaliumdivetyortofosfaatti
Kaava: KH₂PO₄
Kiehumispiste: ∼100 °C (1013 hPa)
Sulamispiste: 253 °C
Tiheys: 2,3 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011401
CAS-numero: 7778-77-0
EINECS: 231-913-4
REACH: 01-2119490224-41
Merck Index: 13,07744

Order Now

Testitulosten erittely

Solubility 222 g/l (20 °C)
Melting Point 253 °C (decomposition)
Molar Mass 136.08 g/mol
Bulk Density 1200 kg/m3
Density 2.34 g/cm3 (20 °C)
pH 4.4 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR