Myo-inositoli 97.0-102.0% (by HPLC on dried basis), EMPROVE® ESSENTIAL Ph. Eur., FCC, NF, SAFC®

Toimittaja: MERCK PRODUCTION CHEMICALS

1.04731.1000 1.04731.9029
1.04731.1000EA 972 EUR
1.04731.1000 1.04731.9029
Myo-inositoli 97.0-102.0% (by HPLC on dried basis), EMPROVE® ESSENTIAL Ph. Eur., FCC, NF, SAFC®
Myo-inositoli

Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).
Corresponds to Ph Eur, NF, FCC

Kaava: C₆H₁₂O₆
Kiehumispiste: 291 °C (1013 hPa)
Sulamispiste: 210 °C
Tiheys: 1,752 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00077932
CAS-numero: 87-89-8
EINECS: 201-781-2
Merck Index: 13,05001

Order Now

Testitulosten erittely

Assay (HPLC, calc.on anhydrous substance) 97.0 - 102.0 %
Identity conforms
Appearance of solution (10 %; water) passes test
Electrical conductivity ≤ 20 µS/cm
Melting range (lower value) ≥ 224 °C
Melting range (upper value) ≤ 227 °C
Chloride (Cl) ≤ 0.0050 %
Sulfate (SO₄) ≤ 0.0050 %
Ba (Barium) conforms
Ca (Calcium) conforms
Pb (Lead) ≤ 0.00005 %
Residual solvents (ICH Q3C) excluded by manufacturing process
Related substances/HPLC (EP) (Impurity A) ≤ 0.3 %
Related substances/HPLC (EP) (Impurity B) ≤ 0.3 %
Related substances/HPLC (NF) (Major unspecified impurity) ≤ 0.3 %
Related substances/HPLC (EP) (Major unspecified impurity) ≤ 0.10 %
Related substances/HPLC (EP) (Sum of all impurities) ≤ 1.0 %
Related substances/HPLC (NF) (Sum of all impurities) ≤ 1.0 %
Sulfated ash ≤ 0.1 %
Loss on Drying (105 °C) ≤ 0.5 %
Water (according to Karl Fischer) ≤ 0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR