Elohopea(II)sulfaatti, extra pure, Supelco®

Toimittaja: Merck
Vaara


M-Clarity™ quality level = MQ200

1.04481.0100 1.04481.0250 1.04481.1000
1.04481.0100EA 258 EUR
1.04481.0100 1.04481.0250 1.04481.1000
Elohopea(II)sulfaatti, extra pure, Supelco®
Elohopea(II)sulfaatti
Kaava: HgSO₄
Sulamispiste: 850 °C
Tiheys: 6,47 g/cm³ (25 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011047
CAS-numero: 7783-35-9
EINECS: 231-992-5
UN: 1645
ADR: 6.1,II
Merck Index: 13,05914

Order Now

Testitulosten erittely

Identity passes test
Chloride (Cl) ≤ 0.003 %
Fe (Iron) ≤ 0.005 %
Mercury (I) (as Hg) ≤ 0.1 %
Substances not reducing by formic acid (residue on ignition) ≤ 0.1 %
Ignition Point 450 °C
Hazard Class 6.1
Hazard Class Description Toxic materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR