Elohopeadikloridi ACS, Reag. Ph. Eur. for analysis, Supelco®

Toimittaja: Merck
Vaara


M-Clarity™ quality level = MQ300

1.04419.0050 1.04419.0250 1.04419.1000
1.04419.0050EA 154 EUR
1.04419.0050 1.04419.0250 1.04419.1000
Elohopeadikloridi ACS, Reag. Ph. Eur. for analysis, Supelco®
Elohopeadikloridi
Kaava: HgCl₂
Kiehumispiste: 302 °C (1013 hPa)
Sulamispiste: 273 °C
Tiheys: 5,44 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011041
CAS-numero: 7487-94-7
EINECS: 231-299-8
UN: 1624
ADR: 6.1,II
Merck Index: 13,05901

Order Now

Testitulosten erittely

Assay (complexometric) ≥ 99.5 %
Assay (complexometric; calculated on dried substance) 99.5 - 100.5 %
Identity passes test
Appearance of solution passes test
Solution in Ether passes test
Acidity or alkalinity passes test
Mercury(I)-chloride passes test
Fe (Iron) ≤ 0.002 %
Residue after reduction ≤ 0.02 %
Loss on drying (24 h im Vakuum) ≤ 1.0 %
Correspons to Reag. Ph Eur,ACS


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR