n-Heptaani ≥99%, EMSURE® Reag. Ph. Eur. for analysis, Supelco®

Toimittaja: Merck
Vaara

Synonyms: high purity solvents

M-Clarity™ quality level = MQ300

1.04379.1000 1.04379.2500 1.04379.6010 1.04379.6025 1.04379.6026 1.04379.2511 1.04379.9910 1.04379.9920 1.04379.4000
1.04379.1000EA 67.5 EUR
1.04379.1000 1.04379.2500 1.04379.6010 1.04379.6025 1.04379.6026 1.04379.2511 1.04379.9910 1.04379.9920 1.04379.4000
n-Heptaani ≥99%, EMSURE® Reag. Ph. Eur. for analysis, Supelco®
n-Heptaani

EMSURE® grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE® provides worldwide best and most extensive product specifications. We declare our EMSURE® range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.

Kaava: H₃C(CH₂)₅CH₃
Kiehumispiste: 98,4 °C (1013 hPa)
Sulamispiste: –90,6 °C
Tiheys: 0,6838 g/cm³ (20 °C)
Leimahduspiste: –4 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00009544
CAS-numero: 142-82-5
EINECS: 205-563-8
UN: 1206
ADR: 3,II
Merck Index: 13,04679
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ff97126664c92362bf0aa2645719548c99f0533&vid=6722523a326fdf1ebabd08a2b8656dd57300787f&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Purity (GC) ≥ 99 %
Identity (IR) conforms
Color ≤ 10 Hazen
Free acid (as CH₃COOH) ≤ 0.0005 %
Density (d 20 °C/20 °C) 0.683 - 0.686
Refractive index (n 20/D) 1.387 - 1.388
Boiling range (97°C-98°C) ≥ 95 % (v/v)
Aromatic hydrocarbons (as C₆H₆) ≤ 0.1 %
Sulfur compounds (as S) ≤ 0.005 %
Readily carbonizable substances conforms
Al (Aluminium) ≤ 0.00001 %
B (Boron) ≤ 0.000002 %
Ba (Barium) ≤ 0.00001 %
Ca (Calcium) ≤ 0.00005 %
Cd (Cadmium) ≤ 0.000005 %
Co (Cobalt) ≤ 0.000002 %
Cr (Chromium) ≤ 0.000002 %
Cu (Copper) ≤ 0.000002 %
Fe (Iron) ≤ 0.00001 %
Mg (Magnesium) ≤ 0.00001 %
Mn (Manganese) ≤ 0.000002 %
Ni (Nickel) ≤ 0.000002 %
Pb (Lead) ≤ 0.00001 %
Sn (Tin) ≤ 0.00001 %
Zn (Zinc) ≤ 0.00001 %
Evaporation residue ≤ 0.001 %
Water ≤ 0.01 %
Ph Eur-reagent


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR