Folic acid 97.0-102.0% (by HPLC) biokemiaan, Millipore®

Toimittaja: Merck


M-Clarity™ quality level = MQ100

1.03984.0005 1.03984.0025 1.03984.1000
1.03984.0005EA 70.9 EUR
1.03984.0005 1.03984.0025 1.03984.1000
Folic acid 97.0-102.0% (by HPLC) biokemiaan, Millipore®
Folic acid
Kaava: C₁₉H₁₉N₇O₆
Sulamispiste: 250 °C
Tiheys: 110 kg/m³
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00079305
CAS-numero: 59-30-3
EINECS: 200-419-0

Order Now

Testitulosten erittely

Assay (HPLC, Calc.on anhydrous substance) 97.0 - 102.0%
Identity (HPLC) Conforms
Identity (IR-spectrum) Conforms
Absorption ratio (A₂₅₆/A₃₆₅) 2.80 - 3.00
Spec. rotation (α 20/D; 10 g/l; NaOH 0,1 mol/l; calc. on anhydrous subst.) +18 - +22°
Related substances (HPLC) (N-(4-aminobenzoyl)-L-glutamic acid (impurity A)) ≤0.5%
Related substances (HPLC) (Pteroic Acid (Impurity D)) ≤0.6%
Related Compounds (HPLC) (Total unspecified impurities) ≤1.0%
Water (According to Karl Fischer) 5.0 - 8.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR