HPLC adapters for solvent supply and waste solvent
Toimittaja: Merck
1.01111.0001 1.09996.0001 1.03833.0001 1.03830.0001 1.03831.0001
1.01111.0001EA 252 EUR
1.01111.0001 1.09996.0001 1.03833.0001 1.03830.0001 1.03831.0001
HPLC adapters for solvent supply and waste solvent
Tulpat ja korkit Turvakorkit
Specially developed screw caps for direct instrument connection of Merck S 40 threaded bottles ensure safe handling of solvents in the lab and protect human health, the environment, as well as solvent quality.