Rauta, EMSURE® 10 µm for analysis, reduced, Supelco®

Toimittaja: Merck
Varoitus


M-Clarity™ quality level = MQ300

1.03819.0100 1.03819.0500
1.03819.0100EA 47.4 EUR
1.03819.0100 1.03819.0500
Rauta, EMSURE® 10 µm for analysis, reduced, Supelco®
Rauta
Kaava: Fe
Kiehumispiste: 3000 °C/1013 hPa
Sulamispiste: 1535 °C
Tiheys: 7,87 g/cm3 (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00010999
CAS-numero: 7439-89-6
EINECS: 231-096-4
UN: 3089
ADR: 4.1,III

Order Now

Testitulosten erittely

Assay (cerimetric) ≥ 99.5 %
Substances insoluble in sulfuric acid ≤ 0.1 %
Chloride (Cl) ≤ 0.002 %
Sulphide (S) ≤ 0.002 %
As (Arsenic) ≤ 0.0002 %
Pb (Lead) ≤ 0.002 %
Cu (Copper) ≤ 0.002 %
Mn (Manganese) ≤ 0.002 %
Zn (Zinc) ≤ 0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR