Dimetyylisulfoksidi, EMSURE® ACS for analysis, Supelco®

Toimittaja: Merck


M-Clarity™ quality level = MQ300

1.02952.1000 1.02952.2500 1.02952.9025 1.02952.2511 1.02952.1011 1.02952.4000
1.02952.1000EA 123 EUR
1.02952.1000 1.02952.2500 1.02952.9025 1.02952.2511 1.02952.1011 1.02952.4000
Dimetyylisulfoksidi, EMSURE® ACS for analysis, Supelco®
Dimetyylisulfoksidi

EMSURE® grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE® provides worldwide best and most extensive product specifications. We declare our EMSURE® range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.

Kaava: (CH₃)₂SO
Kiehumispiste: 189 °C (1013 hPa)
Sulamispiste: 18,5 °C
Tiheys: 1,101 g/cm³ (20 °C)
Leimahduspiste: 87 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00002089
CAS-numero: 67-68-5
EINECS: 200-664-3
Merck Index: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Solubility 1000 g/l (20 °C)
Melting Point 18.4 °C
Molar Mass 78.13 g/mol
Boiling Point 189 °C (1013 hPa)
Vapor Pressure 0.6 hPa (20 °C)
Flash Point 87 °C
Refractive Index 1.48 (20 °C)
Explosion Limit 1.8 - 63.0 %(V)
Density 1.10 g/cm3 (20 °C)
Ignition Point 300 - 302 °C
Saturation concentration 8.0 g/m3 (20 °C)
Viscosity kinematic 2.14 mm2/s


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR