Kupari(II)sulfaatti-5 H2O, crystallised, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, SAFC®

Toimittaja: MERCK PRODUCTION CHEMICALS
Vaara

1.02787.1000 1.02787.5000 1.02787.9050
1.02787.1000EA 472 EUR
1.02787.1000 1.02787.5000 1.02787.9050
Kupari(II)sulfaatti-5 H2O, crystallised, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, SAFC®
Kupari(II)sulfaatti-5 H2O
Kaava: CuSO₄·5H₂O
Tiheys: 2,284 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149681
CAS-numero: 7758-99-8
EINECS: 231-847-6
UN: 3077
ADR: 9,III
REACH: 01-2119520566-40
Merck Index: 13,02682

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR