Kalsiumasetaatti-xH2O DAC, FCC, extra pure, SAFC®

Toimittaja: MERCK PRODUCTION CHEMICALS

1.02052.1000 1.02052.9029
1.02052.1000EA 239 EUR
1.02052.1000 1.02052.9029
Kalsiumasetaatti-xH2O DAC, FCC, extra pure, SAFC®
Kalsiumasetaatti-xH2O
Kaava: (H₃CCOO)₂Ca·nH₂O
Tiheys: 1,5 g/cm³ (20 °C)
MDL numero: MFCD00150019
CAS-numero: 114460-21-8
EINECS: 200-540-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR