Kalsiumhydroksidi, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®

Toimittaja: Merck
Vaara


M-Clarity™ quality level = MQ300

1.02047.0500 1.02047.1000 1.02047.9050
1.02047.0500EA 61.7 EUR
1.02047.0500 1.02047.1000 1.02047.9050
Kalsiumhydroksidi, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Kalsiumhydroksidi
Kaava: Ca(OH)₂
Sulamispiste: 550 °C
Tiheys: 2,24 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00010901
CAS-numero: 1305-62-0
EINECS: 215-137-3
UN: 3262
ADR: 8,III
Merck Index: 13,01675

Order Now

Testitulosten erittely

Solubility 1.7 g/l (20 °C)
Melting Point 550 °C (decomposition)
Molar Mass 74.09 g/mol
Bulk Density 400 kg/m3
Boiling Point 2850 °C (1013 hPa) (decomposition)
Vapor Pressure (20 °C)
Density 2.24 g/cm3 (20 °C)
pH 12.6 (H2O, 20 °C) (saturated solution)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR