1-Butanoli, LiChrosolv® HPLC, Supelco®

Toimittaja: Merck
Vaara

Synonyms: 1-Butyl alcohol, Butyl Alcohol, 1-Hydroxy butane, Propyl carbinol, n-Butanol, n-Butyl Alcohol

MQ100

1.01988.1000 1.01988.2500
1.01988.1000EA 207 EUR
1.01988.1000 1.01988.2500
1-Butanoli, LiChrosolv® HPLC, Supelco®
1-Butanoli
Kaava: H₃C(CH₂)₂CH₂OH
MW: 74.12 g/mol
Kiehumispiste: 117.6
Sulamispiste: –89 °C
Tiheys: 0.812 g/cm³ (20 °C)
Leimahduspiste: 30
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00002964
CAS-numero: 71-36-3
EINECS: 200-751-6
UN: 1120
ADR: 3 III
REACH: 01-2119484630-38
Merck Index: 13,01539
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9395eeb6150a74222da52460f53cedeccd93442&vid=fd8352a556914c5be7213c576355a88e130d1a7b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR