Petrolieetteri, 80…100 °C, EMSURE® analysis_for, Supelco®

Toimittaja: Merck
Vaara

Synonyms: Gasoline, Petroleum ether, Petrol, Benzine

MQ300

1.01777.1000
1.01777.1000EA 71.1 EUR
1.01777.1000
Petrolieetteri, 80…100 °C, EMSURE® analysis_for, Supelco®
Petrolieetteri
Kiehumispiste: 80…100 °C (1013 hPa)
Tiheys: 0.69 g/cm³ (20 °C)
Leimahduspiste: –10 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00081849
CAS-numero: 64742-49-0
EINECS: 265-151-9
UN: 1268
ADR: 3 II

Order Now

Testitulosten erittely

Solubility (25 °C) insoluble
Boiling Point 80 - 100 °C (1013 hPa)
Vapor Pressure 60 hPa (20 °C)
Explosion Limit 1.0 - 6.5 %(V)
Density 0.69 g/cm3 (20 °C)
Ignition Point 250 °C
Viscosity kinematic 0.6 mm2/s (20 °C)

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR