Petrolieetteri, 40…80 °C for denaturation, Supelco®

Toimittaja: Merck
Vaara

Synonyms: Gasoline, Petroleum ether, Petrol, Benzine

M-Clarity™ quality level = MQ200

1.01769.1000 1.01769.6010 1.01769.6025 1.01769.9190
1.01769.1000EA 76.9 EUR
1.01769.1000 1.01769.6010 1.01769.6025 1.01769.9190
Petrolieetteri, 40…80 °C for denaturation, Supelco®
Petrolieetteri
Kiehumispiste: 40…70 °C (1013 hPa)
Tiheys: 0,666 g/cm³ (15 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00081849
CAS-numero: 64742-49-0
EINECS: 265-151-9
UN: 1268
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR