Typpihappo 60%, Ultrapure for trace analysis, Supelco®

Toimittaja: Merck
Vaara

Synonyms: Nitric acid solution

M-Clarity™ quality level = MQ100

1.01518.0250 1.01518.0500 1.01518.1000
1.01518.0250EA 711 EUR
1.01518.0250 1.01518.0500 1.01518.1000
Typpihappo 60%, Ultrapure for trace analysis, Supelco®
Typpihappo
Kaava: HNO₃
Kiehumispiste: ∼120 °C (1013 hPa)
Sulamispiste: –22 °C
Tiheys: 1,37 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011349
CAS-numero: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8,II

Order Now

Testitulosten erittely

Solubility (20 °C) soluble
Melting Point -22 °C
Boiling Point 120 °C (1013 hPa)
Density 1.37 g/cm3 (20 °C)
pH  - 1 (H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR