Testipakkaukset, AOX, tuotteille Spectroquant®, Supelco®
Toimittaja: Merck
1.00675.0001 1.00677.0001 1.00678.0001 1.00680.0001
1.00675.0001EA 255 EUR
1.00675.0001 1.00677.0001 1.00678.0001 1.00680.0001
Testipakkaukset, AOX, tuotteille Spectroquant®, Supelco®
Ympäristöanalytiikan pikatestit
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells for most the test parameters
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
*1 AOX Enrichment Set 1.00678.0001 and AOX Sample Preparation Set 1.00677.0001 required.