Tungstosilicic acid hydrate, EMSURE® for analysis, Supelco®

Toimittaja: Merck


M-Clarity™ quality level = MQ300

1.00659.0100
1.00659.0100EA 194 EUR
1.00659.0100
Tungstosilicic acid hydrate, EMSURE® for analysis, Supelco®
Tungstosilicic acid Hydrate
Kaava: H₄Si(W₃O₁₀)₄.xH₂O
MDL numero: MFCD00036271
CAS-numero: 12027-43-9
EINECS: 234-719-8

Order Now

Testitulosten erittely

Solubility (20 °C) soluble
Molar Mass 2878.17 g/mol
Bulk Density 200 - 500 kg/m3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR