N,N-Dimetyyliformamidi for peptide synthesis, Supelco®

Toimittaja: Merck
Vaara


M-Clarity™ quality level = MQ200

1.00397.2500 1.00397.6025 1.00397.4000 1.00397.6190
1.00397.2500EA 283 EUR
1.00397.2500 1.00397.6025 1.00397.4000 1.00397.6190
N,N-Dimetyyliformamidi for peptide synthesis, Supelco®
N,N-Dimetyyliformamidi
Kaava: HCON(CH₃)₂
Kiehumispiste: 153 °C (1013 hPa)
Sulamispiste: –61 °C
Tiheys: 0,949 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003284
CAS-numero: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3,III
REACH: 01-2119475605-32
Merck Index: 13,03269
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58f54ca04f44541371e8800ec9787397bc18cfbe&vid=2a2880fd49e90c177da16050a7481943082095e8&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Purity (GC) ≥ 99.9 %
Identity (IR) conforms
Color ≤ 10 Hazen
Acidity ≤ 0.0005 meq/g
Alkalinity ≤ 0.0002 meq/g
Amines (Dimethylamine) ≤ 10 ppm
Evaporation residue ≤ 10
Water ≤ 300 ppm
Density 0.94 g/cm3 (20 °C)
pH 7 (200 g/l, H2O, 20 °C)
Ignition Point 410 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR