Bromivety 47%, Supelco®

Toimittaja: Merck
Vaara


M-Clarity™ quality level = MQ200

1.00304.0500 1.00304.2500 1.00304.9020
1.00304.0500EA 147 EUR
1.00304.0500 1.00304.2500 1.00304.9020
Bromivety 47%, Supelco®
Bromivety
Kaava: HBr
Tiheys: 1,47 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011323
CAS-numero: 10035-10-6
EINECS: 233-113-0
UN: 1788
ADR: 8,II

Order Now

Testitulosten erittely

Solubility (20 °C) soluble
Melting Point -11 °C
Boiling Point 126 °C (1013 hPa)
Vapor Pressure 10.6 hPa (20 °C)
Density 1.49 g/cm3 (20 °C)
pH (H2O, 20 °C) strongly acid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR