L(+)-glutamiinihappo, Millipore®

Toimittaja: Merck


M-Clarity™ quality level = MQ100

1.00291.0250 1.00291.1000 1.00291.9010
1.00291.0250EA 93.9 EUR
1.00291.0250 1.00291.1000 1.00291.9010
L(+)-glutamiinihappo, Millipore®
L(+)-glutamiinihappo

Soluble in water and mineral acids; insoluble in organic solvents. In aqueous solutions forms pyrrolidonecarboxylic acid slowly at room temperature, more rapidly at 100 °C.

Kaava: HO₂CCH₂CH₂CH(NH₂)CO₂H
Kiehumispiste: 325 °C (1013 hPa)
Sulamispiste: 160 °C
Tiheys: 1,538 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00002634
CAS-numero: 56-86-0
EINECS: 200-293-7
Merck Index: 13,04482
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1cb2faefcc53483f0b3bfccd6c12b611e487241&vid=a462989da97f7186f34fef7de9cad6596c2589ee&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR