L(+)-glutamiini, Millipore®

Toimittaja: Merck

Synonyms: Gln, L-Glutamine, H-Gln-OH, (S)-(+)-Glutamine, (S)-2,5-Diamino-5-oxopentanoic acid, Levoglutamide, Q

M-Clarity™ quality level = MQ100

1.00289.1000 1.00289.9010
1.00289.1000EA 505 EUR
1.00289.1000 1.00289.9010
L(+)-glutamiini, Millipore®
L(+)-glutamiini
Kaava: C₅H₁₀N₂O₃
Kiehumispiste: 355 °C (1013 hPa)
Sulamispiste: 185…186 °C
Tiheys: 1,5326 g/cm³ (130 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00008044
CAS-numero: 56-85-9
EINECS: 200-292-1
Merck Index: 13,04484
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb7a812ec5d2c73a797fc464926df9eef12c5c9a&vid=15d7d87daff0fec33a974a578415e157a3278049&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR